Two Way Valves
Two Way Valves
Actual product may vary
Fisherbrand
Two Way Valves
Two Way Valves are used for transfer of liquids or gases in order to release the flow, to block it or to change its direction.
Have Questions?
Catalog NumberMaterialFor Use WithOverall Width (Metric)
FKS10180811Polypropylene, Polyethylene9 to 11 mm dia. Hose8 mm
Catalog number FKS10180811
Price (SGD)
42.00
EA
Add to cart
Material:
Polypropylene, Polyethylene
For Use With:
9 to 11 mm dia. Hose
Overall Width (Metric):
8 mm
Price (SGD)
42.00
EA
Add to cart
  • For transfer of liquids or gases in order to release the flow, to block it or to change its direction
  • PVDF: excellent chemical resistance, transparent, suitable for food
  • PP/PE: for industrial use, red/white
  • Operating temperature: PVDF +0 up to +40 °C, PP/PE +5 up to +40 °C
  • Resistant to compression max. 1 bar
  • With mounting plate for easy installation
Specifications
For Use With9 to 11 mm dia. Hose
IncludesMounting Plate
Max. Operating Temperature (Metric)40°C
MaterialPolypropylene, Polyethylene
Min. Operating Temperature (Metric)5°C
Overall Width (Metric)8 mm
Type2-way Valve
Unit SizeEA