Combine these replacement microcentrifuge rotor lids with the microcentrifuge rotor.
Have Questions?
Change view
Catalog Number
Type
FKS10177222
ClickSeal Lid
75003410
ClickSeal Lid
2 Options
Catalog number FKS10177222
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Type:
ClickSeal Lid
Combine Thermo Scientific™ Replacement Microcentrifuge Rotor Lids with the microcentrifuge rotor.
ClickSeal™ Biocontainment Lid:
For use with 24 × 1.5/2.0 mL Rotor (75003424), PCR 4 × 8 Rotor (75003440)
Unique design clicks when proper seal is made, providing assurance that your samples are securely contained, as well as shortens retrieval time with one click opening
With PPSU rotor lid, view contents of rotor for added safety and convenience
Screw-on Lid:
For use with 36 × 0.5 mL Rotor (75003436), Dual Row 18 × 2.00.5 mL Rotor (75003418), PCR 8 × 8 Rotor (75003489)
Ordering Information:
Sold individually.
Specifications
TypeClickSeal Lid
For Use With (Equipment)24 x 1.5/2 mL Rotor (75003424), PCR 4 x 8 Rotor