Check Order Status
Silicone Pump Tubing
Actual product may vary
Fisherbrand
Silicone Pump Tubing
For use in field of medicine.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Inner
Thickness (Metric)
FKS10172031
4.8 mm
2.4 mm
FKS10162031
1.6 mm
2.4 mm
FKS10010061
6.4 mm
2.4 mm
3 Options
Catalog number
FKS10172031
Price (SGD)
520.00
EA
Add to cart
Diameter (Metric) Inner:
4.8 mm
Thickness (Metric):
2.4 mm
Price (SGD)
520.00
EA
Add to cart
FDA, NWC and BGA approved
Certified to USP XXII class 6
Autoclavable at 130°C in steam
Sterilizable by gamma radiation without impairing quality
Maybe opaque or translucent in appearance
Specifications
Certifications/Compliance
Class 6
Autoclavable
Autoclavable
Diameter (Metric) Inner
4.8 mm
Material
Silicone
Overall Length (Metric)
1500 cm
Thickness (Metric)
2.4 mm
For Use With
Peristaltic Pump
Packaging Quantity
1 Each
Color
Translucent
Product Type
Pump Tubing