ART™ Barrier Specialty Pipette Tips
ART™ Barrier Specialty Pipette Tips
Actual product may vary
ART™ Barrier Specialty Pipette Tips
ART™ Barrier Specialty Pipette Tips
ART™ Barrier Specialty Pipette Tips
ART™ Barrier Specialty Pipette Tips
ART™ Barrier Specialty Pipette Tips
Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FKS10170621180 μL
2159P180 μL
FIS21-236-2B180 μL
2279B1000 μL
Catalog number FKS10170621
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
180 μL
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
Designed to aid in pipetteting for a variety of applications:
Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading for loading acrylamide or agarose gels
Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials
Solvent Safe for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettetes
Ergonomic pipette tips are designed for Gilson pipetteman™ pipettetes and Thermo Scientific™ Finnpipettete™ Focus pipettetes
Specifications
DescriptionART XLPSterile,
Volume (Metric)180 μL
FilteredFiltered
SterilitySterile
ModelART XLP
Length (English)3.59 in.
Length (Metric)9.12cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatRack
Tip StyleExtended Length
For Use With (Equipment)Gilson™ Pipetman™, Thermo Scientific™ Finnpipette™ Focus
Product LineART
Fit for PurposeSterile and PCR Ready
Unit SizePK