Tygon™ E-3603 Tubing
Tygon™ E-3603 Tubing
Actual product may vary
Tygon™ E-3603 Tubing
Tygon™ E-3603 Tubing
Fisherbrand
Tygon™ E-3603 Tubing
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ E-3603 Tubing handles virtually all inorganic chemicals found in the lab.
Have Questions?
Catalog NumberDiameter (Metric) InnerDiameter (Metric) OuterThickness (Metric)Overall Length (Metric)
FKS101701614 mm7.1 mm1.6 mm15 m
Catalog number FKS10170161
Price (SGD)
352.00
EA
Add to cart
Diameter (Metric) Inner:
4 mm
Diameter (Metric) Outer:
7.1 mm
Thickness (Metric):
1.6 mm
Overall Length (Metric):
15 m
Price (SGD)
352.00
EA
Add to cart
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Performance Plastics™ Tygon E-3603 Tubing handles virtually all inorganic chemicals found in the lab.
Tygon™ E-3603 tubing from Saint-Gobain is crystal clear and flexible, handling virtually all organic chemicals found in the modern laboratory. It is non-oxidizing and non-contaminating, making it an ideal choice for a variety of applications. Tygon™ E-3603 has high tolerances and lot-to-lot consistency, which will yield reproducible results in your laboratory. It is non-toxic and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning.
Tygon™ E-3603 tubing is specially formulated for resistance to flex-fatigue and abrasion. As a tubing for instrumentation connection, vent, drain and other general laboratory applications, Tygon™ E-3603 tubing offers superior life, which minimizes the labor and the expenses associated with replacement.
Specifications
Certifications/ComplianceFDA, NSF, 3-A, USP Class VI, Japan Food Sanitation Law #370/1959, 1935/2004/EC, 10/2011/EU
AutoclavableAutoclavable
Diameter (English) Inner0.16 in.
Diameter (Metric) Inner4 mm
MaterialTygon E-3603
Diameter (English) Outer0.28 in.
Diameter (Metric) Outer7.1 mm
Overall Length (English)49.2 ft.
Overall Length (Metric)15 m
Thickness (English)0.06 in.
Thickness (Metric)1.6 mm
For Use With (Application)For laboratory, food and, beverage and vacuum applications
Packaging Quantity1 Ea.
ColorClear
FlexibilityFlexible
Maximum Pressure2.5 bar (at 23°C)
Vacuum Level760 mm Hg (at 23°C)
Max. Operating Temperature (Metric)74°C
Product TypeFlexible Tubing
Unit SizeEA