Volac™ Glass Pasteur Pipettes
Volac™ Glass Pasteur Pipettes
Actual product may vary
Fisherbrand
Volac™ Glass Pasteur Pipettes
Ideal for transferring samples to sample cups and for drop counting. Poulten Graf™ Volac™ Glass Pasteur Pipettes are pre-plugged and feature a heavier wall thickness to reduce the risk of breakage in use or when fitting PVC teats.
Have Questions?
Catalog NumberType
FKS10167901Pasteur Pipet
Catalog number FKS10167901
Price (SGD)
808.00
PK
Add to cart
Type:
Pasteur Pipet
Price (SGD)
808.00
PK
Add to cart
  • Nominal rate of delivery 50 drops/mL
  • Precision made on fully automatic machines to ensure total uniformity
  • Constrictions are heat formed to avoid weakness or contamination of glass
Specifications
Certifications/ComplianceISO7712
MaterialGlass
IncludesCotton Plug
Quantity1000/Pk.
Length (Metric)150 mm
TypePasteur Pipet
Unit SizePK