MASTER-20T Refractometer
MASTER-20T Refractometer
Actual product may vary
Fisherbrand
MASTER-20T Refractometer
Easy-to-read, easy-to-hold, and smooth-sampling refractometer. Atago™ MASTER-20T Refractometer is a low-concentration model.
Have Questions?
Catalog NumberScaleRange
FKS10166374
Catalog number FKS10166374
Price (SGD)
666.00
EA
Add to cart
Price (SGD)
666.00
EA
Add to cart
  • Low concentration
  • Easy disposal of sample from the edge
  • Textured grip
  • Well balanced
  • Streamlined body
  • Easily cleaned with running water
  • Convenient storage case
  • Automatic sample distribution
  • Sample size: 2-3 drops
  • Automatic temperature compensation
Specifications
TypeRefractometer
Scale Range0 to 20 Brix
ModelMASTER-20T
Accuracy±0.2% Brix 10 to 30°C
Repeatability±0.1% Brix
Scale TypeBrix
For Use With (Application)Measurements of low concentration fruit juices, cutting oil
Weight (Metric)165 g
Dimensions (L x W x H)20.7 x 3.2 x 3.4 cm
Unit SizeEA