Whatman™ Grade 54 Quantitative Filter Paper
Whatman™ Grade 54 Quantitative Filter Paper
Actual product may vary
Fisherbrand
Whatman™ Grade 54 Quantitative Filter Paper
Supports collection of coarse and gelatinous precipitates with minimal fiber shredding, even when wet. Whatman™ Grade 54 Quantitative Filter Paper has a hard, smooth surface to sustain vacuum and suction filtration.
Have Questions?
Catalog NumberFormatDiameter (Metric)DimensionsLxW
FKS10163971Disc150 mm
Catalog number FKS10163971
Price (SGD)
308.00
PK
Add to cart
Format:
Disc
Diameter (Metric):
150 mm
Price (SGD)
308.00
PK
Add to cart
Supports collection of coarse and gelatinous precipitates with minimal fiber shredding, even when wet. Whatman™ Grade 54 Quantitative Filter Paper has a hard, smooth surface to sustain vacuum and suction filtration.
  • Acid-treated for high wet-strength and chemical resistance
  • Large nominal particle retention rating of 22μm
  • Fast filtration speed (Herzberg filtration speed 39 s)
  • High purity (0.015% ash maximum)
  • Compatible with Büchner funnel or Hirsch funnel applications
Specifications
Airflow Rate3 s/100 mL/in2
DescriptionGrade 54 Filter for Quantitative Analysis, Fast, 150 mm circle (100 pcs)
Filter TypeHardened Low Ash
FormatDisc
MaterialCotton Linters
Particle Retention22 μm
Pore Size22 μm
Filtration SpeedASTM, 10 seconds; Herzberg, 39 seconds
Flow RateFast
GradeGrade 54
PorosityCoarse
Quantity100 Pk.
Thickness (Metric)185 μm
TypeQuantitative Filter Paper
Diameter (English)5.9 in.
Diameter (Metric)150 mm
Unit SizePK