Baffled Polycarbonate Erlenmeyer Flasks with Cap
Baffled Polycarbonate Erlenmeyer Flasks with Cap
Actual product may vary
Fisherbrand
Baffled Polycarbonate Erlenmeyer Flasks with Cap
Designed with baffles to provide premium agitation at lower speed for gentler, more efficient aeration. Corning™ Baffled Polycarbonate Flasks with Caps are ideal for shaker culture applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure TypeDiameter (Metric) ClosureGraduations
FKS10160253250 mLVented38 mm25 mL
FIS13-700-429125 mLVented33 mm25 mL
FIS13-700-4261 LFlat43 mm50 mL
FIS13-700-431250 mLVented38 mm25 mL
Catalog number FKS10160253
Price (SGD)
2,750.00
PK
Add to cart
Capacity (Metric):
250 mL
Closure Type:
Vented
Diameter (Metric) Closure:
38 mm
Graduations:
25 mL
Price (SGD)
2,750.00
PK
Add to cart
Designed with baffles to provide premium agitation at lower speed for gentler, more efficient aeration. Corning™ Baffled Polycarbonate Flasks with Caps are ideal for shaker culture applications.
  • Made from optically clear polycarbonate
  • Ideal for shaker culture applications
  • Sterilized by gamma radiation and nonpyrogenic
  • RNase- and DNase-free
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
Capacity (English)8.45 oz.
Closure ColorOrange
Closure MaterialPolypropylene
DescriptionTop style: Threaded
Diameter (Metric) Closure38 mm
Diameter (Metric) Outer Neck3.1 cm
Capacity (Metric)250 mL
MaterialPolycarbonate
For Use With (Application)For shaker culture applications
Graduations25 mL
Neck StyleWide Mouth
No. per Case50/Cs.
No. per Pack1/Pk.
DisposableSingle-use
Mouth StyleWide
SterilitySterile
Bottom ShapeBaffled
Closure TypeVented
Closure Size38 mm
Unit SizePK