Quantofix™ Peroxide 100 Test Strip Kits
Quantofix™ Peroxide 100 Test Strip Kits
Actual product may vary
Fisherbrand
Quantofix™ Peroxide 100 Test Strip Kits
Allows for the quick and easy determination of peroxide levels in solutions. Macherey Nagel™ Quantofix™ Peroxide 100 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Have Questions?
Catalog NumberColor Indicator
FKS10159960White to Blue
Catalog number FKS10159960
Price (SGD)
152.00
PK
Add to cart
Color Indicator:
White to Blue
Price (SGD)
152.00
PK
Add to cart
Allows for the quick and easy determination of peroxide levels in solutions. Macherey Nagel™ Quantofix™ Peroxide 100 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
  • Ready to use
  • Rapid: provides reliable results within 15 seconds
  • CE marked and suitable to be used in the healthcare environment
  • Can be used in chemical laboratories to check organic solvents
  • Test for residual peroxide in cleaning procedures with high potency disinfectants
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store at room temperature
Specifications
Color IndicatorWhite to Blue
FormatStrip
For Use With (Application)Determination of Peroxide 100
Graduations0, 1, 3, 10, 30, 100mg/L H2O2
Quantity100 Pack
Shelf Life2.5 Years
Storage RequirementsRoom Temperature
Test Time15 sec.
TypeSemi-qualitative Test Strip
Unit SizePK