Amicon™ Ultra-4 Centrifugal Filter Units
Amicon™ Ultra-4 Centrifugal Filter Units
Actual product may vary
Fisherbrand
Amicon™ Ultra-4 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
Have Questions?
Catalog NumberMWCOQuantityDiameter (Metric)
FKS1015891110 kDa96 Pack17.3 mm
Catalog number FKS10158911
Price (SGD)
3,496.00
PK
Add to cart
MWCO:
10 kDa
Quantity:
96 Pack
Diameter (Metric):
17.3 mm
Price (SGD)
3,496.00
PK
Add to cart
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange
Specifications
DisposableSingle-use
Material (Membrane)High-recovery Ultracel-PL Regenerated Cellulose
MWCO10 kDa
SterilityNon-sterile
Filtration Area3 cm2
For Use With (Application)Protein purification, protein concentration and desalting
Housing MaterialRegenerated Cellulose (RC)
IncludesTwo microcentrifuge tubes
Quantity96 Pack
TypeCentrifugal Filters
Volume (Metric)4 mL
Length (Metric)124 mm
Diameter (Metric)17.3 mm
Unit SizePK