1x1 image pixel for data collection
General Assay Microplate Lids
General Assay Microplate Lids
Actual product may vary
Fisherbrand
General Assay Microplate Lids
Manufactured with rigid polystyrene for optimum visibility and strength. Corning™ General Assay Microplate Lids are available in a variety of styles and types.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate CompatibilityNo. per CaseMaterial
FKS10157051All Microplates100/Cs.Polystyrene
FIS07-200-59896-well Microplates Only (not 2 mL block)50/Cs.Polystyrene
Catalog number FKS10157051
Price (SGD)
1,220.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Plate Compatibility:
All Microplates
No. per Case:
100/Cs.
Material:
Polystyrene
Price (SGD)
1,220.00
PK
Add to cart
  • All lids are made of rigid polystyrene except where indicated.
  • All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning™ microplates.
  • The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
  • The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
  • The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
  • Specifications
    MaterialPolystyrene
    SterilitySterile
    No. per Case100/Cs.
    No. per Pack25/Pk.
    Product TypeUniversal Lid
    Plate CompatibilityAll Microplates
    Cut-off CornersNo
    Unit SizePK
    Documents & Downloads
    Certificates
    0 results displayed, search above for a specific certificate
    Share catalog number, name or link