Tubes™ 3810X
Tubes™ 3810X
Actual product may vary
Fisherbrand
Tubes™ 3810X
The easy-to-open, securely closing, Eppendorf Tubes™ 3810X offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry. Safer than ever: designed for centrifugation up to 30,000 x g.
Have Questions?
Catalog NumberColorPurity or Quality Grade
FKS10154671ClearPCR Clean
Catalog number FKS10154671
Price (SGD)
281.00
PK
Add to cart
Color:
Clear
Purity or Quality Grade:
PCR Clean
Price (SGD)
281.00
PK
Add to cart
Use these tubes for all applications in molecular biology, clinical chemistry and biochemistry. Eppendorf™ 3810X Polypropylene Microcentrifuge Tube offers comfort and dependability for sample preparation, centrifugation and storage.
  • Tube lid is easy to open and close for ergonomic operation.
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes.
  • g-Safe™: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
  • Graduation for visual control of the pipetted volume.
  • Frosted lid and surface on the side for easy labeling.
  • Ensured functionality from -86°C to 100°C.
  • Autoclavable when open (121°C, 20 min).
  • Available in Eppendorf Quality and lot-certified PCR clean purity grade.
Specifications
AutoclavableAutoclavable
Capacity (Metric)1.5 mL
ColorClear
MaterialPolypropylene
No. per Pack1000 Pk.
Temperature Range (English)-122.8°F to +212°F
Temperature Range (Metric)-86°C to +100°C
SterilityNon-sterile
Purity or Quality GradePCR Clean
GraduatedYes
Capacity (English)0.05 oz.
Closure TypeFlat Frost Lid
ShapeCylindrical
Fit for PurposePCR Ready
Bottom ShapeConical
Max. RCF30,000 x g
Product TypeMicrocentrifuge Tube
Unit SizePK