Class A Volumetric Flasks with Poly Stopper
Class A Volumetric Flasks with Poly Stopper
Actual product may vary
Fisherbrand
Class A Volumetric Flasks with Poly Stopper
Achieve precise standard solutions. Hirschmann™ Class A Volumetric Flasks with Poly Stopper is standardized to contain or to deliver a definite amount of liquid. Flask has silver diffusion ink with brown graduations.
Have Questions?
Catalog NumberCapacity (Metric)Standard Taper Joint
FKS101517425000 mL34/35 NS Standard Ground
Catalog number FKS10151742
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
5000 mL
Standard Taper Joint:
34/35 NS Standard Ground
  • With polyethylene stopper
  • With amber stain graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
CalibrationTo Contain
Certifications/ComplianceDIN EN ISO 1042
Closure MaterialPolyethylene
ColorPermanent amber markings with silver diffusion color
Capacity (Metric)5000 mL
MaterialBorosilicate Glass (3.3)
Height (Metric)470 ± 5 mm
IncludesPolyethylene Stopper
Limit of Error± 1.2 mL
No. per Pack1/Pack
Standard Taper Joint34/35 NS Standard Ground
Tolerance± 1.2 mL
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
SterilityNon-sterile
ClassA
Bottom ShapeRound
Unit SizeEA