Uvette™ Routine Pack
Uvette™ Routine Pack
Actual product may vary
Fisherbrand
Uvette™ Routine Pack
Use this cuvette for superb purity, convenience and precision for photometric analysis. Eppendorf™ Uvette™ Routine Packs are disposable and feature a light transmission of 220 nm to 1,600 nm. Deep-lying optical window prevents scratches.
Have Questions?
Catalog number FKS10145022
Price (SGD)
832.00
PK
Add to cart
Price (SGD)
832.00
PK
Add to cart
Use this cuvette for superb purity, convenience and precision for photometric analysis. Eppendorf™ Uvette™ Routine Packs are disposable and feature a light transmission of 220 nm to 1,600 nm. Deep-lying optical window prevents scratches.
The unique shape of the UVette combines two optical light path lengths to offer greater flexibility. UVettes in routine packs are ideal for routine applications. These packs allow a safe storage of the UVettes in a re-closable box, while offering convenient access to each cuvette for easy filling.
UV transparent plastic, free of fluoropolymers and other halogenated hydrocarbons
Two built-in optical path lengths in a single cuvette - just turn the UVette 90° to change from 10mm and 2mm
Specifications
TypeCuvette Pack
Capacity (Metric)50 to 2000 μL
MaterialUV-transparent Plastic
Quantity200 Pack
Path Length2 mm or 10 mm
Wavelength Range220 to 1600 nm
For Use With (Equipment)Biophotometers and Spectrophotometers
Unit SizePK