Agencourt AMPure XP
Product Image
Agencourt AMPure XP
Trusted and recommended reagent for size selection and cleanup steps within a variety of NGS workflows. Beckman Coulter™ Agencourt AMPure XP removes unwanted contaminants from DNA in a variety of applications including PCR, NGS, cloning, and microarrays.
Have Questions?
Catalog NumberVolume
FKS101362245mL
Catalog number FKS10136224
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume:
5mL
Agencourt AMPure XP is a highly efficient, easily automated PCR purification system that delivers superior quality DNA with no salt carryover. Requiring no centrifugation or filtration, Agencourt AMPure XP can be easily used in manual and automated 96- or 384-well formats.
  • SPRI paramagnetic bead-based chemistry
  • Removes contaminants (dNTPs, salts, primers, primer dimers)
  • High recovery of amplicons (>100 bp) ensures that critical data is not lost
  • Trusted and recommended reagent for a variety of NGS workflows
  • Compatible with manual and automated processing
  • Predictable and consistent size selection
  • Manual cleanups using AMPure XP require less than 30 minutes
  • AMPure XP is automated on all Biomek platforms for higher throughput
Specifications
CompositionAMPure XP, Speed Beads (Mag Carboxy)
FormLiquid
For Use With (Equipment)Automated 96- or 384-well formats, e.g. Biomek platforms
pH8.0 to 8.4
For Use With (Application)Standard PCR, Real Time PCR (qPCR), Next-Generation Sequencing (NGS)
Sample TypeDNA
Volume5mL
Unit SizeEA
Contents & Storage
2-8°C