Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
Actual product may vary
Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
Fisher Catalogue
Wheaton™ Clear Straight-Sided Jars with Polyvinyl-lined Caps
Ideal for soil sampling and environmental applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Packaging
FKS101333421000 mLConvenience pack
FIS02-911-407125 mLConvenience pack
FIS02-911-408250 mLConvenience pack
FIS02-911-4101000 mLConvenience pack
Catalog number FKS10133342
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Packaging:
Convenience pack
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
  • Wide mouths provide easy access to contents when using spoons or spatulas
Each case of convenience pack bottles includes hazard communication labels for proper content identification and tamper evident labels to protect sample integrity in transit or long-term storage
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorClear
Diameter (English)3.74 in.
MaterialGlass
PackagingConvenience pack
Closure ColorWhite
Height (Metric)170 mm
Height (English)6.69 in.
Closure LinerPoly/Vinyl
Closure MaterialPolypropylene
Closure Size89-400 mm
Diameter (Metric)95 mm
Closure TypeScrew Cap
Unit SizePK