IKA Vortex 3 Shaker
IKA Vortex 3 Shaker
Actual product may vary
Fisherbrand
IKA Vortex 3 Shaker
IKA Vortex 3 Shaker suitable for short-time operation (touch function), activated by pressing shaker attachment or continuous operation.
Have Questions?
Catalog number FKS10132562
Price (SGD)
2,572.00
EA
Add to cart
Price (SGD)
2,572.00
EA
Add to cart
  • Compact design
  • Wide speed range, infinitely adjustable
  • Different applications, 3 interchangeable attachments and 7 inserts (e.g. microtubes, microtiter plates, Erlenmeyer flasks 250mL etc.), (order separately)
  • Attachments securely click onto appliance in any position
  • Special strap ensures easy handling of round/Erlenmeyer flasks
  • Sturdy metal zinc die cast casing
  • Short-time operation activated by pressing attachment (touch function)
  • Stable at high speeds due to special feet (silicon base with ultra high vibration damping)
  • Eccentric with ball bearings
  • Suitable for continuous operation with low self heating thanks to self ventilation of motor
  • Shaking movement: orbital
  • Orbital diameter: 4mm
  • Infinitely adjustable speed range: 500 - 2.500rpm
Specifications
Orbit4 mm
Speed Range0 to 2500 rpm
Voltage200/240 V, 100/115 V
Wattage60 W
Frequency50/60 Hz
Depth (English)5.8 in.
Depth (Metric)14.9 cm
Height (English)5.35 in.
Height (Metric)13.6 cm
Width (English)5 in.
Width (Metric)12.7 cm
Weight (English)9.9 lb.
Weight (Metric)4.5 kg
IP RatingIP 21
Operating ModesContinuous
Unit SizeEA