MF-Millipore™ Membrane Filter, 0.05 μm
MF-Millipore™ Membrane Filter, 0.05 μm
Actual product may vary
Fisherbrand
MF-Millipore™ Membrane Filter, 0.05 μm
13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic
Have Questions?
Catalog NumberDiameter (Metric)
FKS1012638047 mm
Catalog number FKS10126380
Price (SGD)
2,487.00
PK
Add to cart
Diameter (Metric):
47 mm
Price (SGD)
2,487.00
PK
Add to cart
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features & Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
AutoclavableNot Autoclavable
ColorWhite
MaterialMCE
Pore Size0.05 μm
SterilityNon-sterile
Flow Rate0.74 mL/min x cm2
Thickness (Metric)105 μm
TypeFilter Membranes
WettabilityHydrophilic
Diameter (Metric)47 mm
Unit SizePK