Check Order Status
Soda Lime Glass Short Stem Funnel
Actual product may vary
Fisherbrand
Soda Lime Glass Short Stem Funnel
Manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Short Stem Funnel is less likely to clog when funneling thicker substances.
Have Questions?
Change view
Catalog Number
Diameter (Metric)Top
Diameter (Metric) Stem
Length (Metric) Stem
FKS10124352
55mm
8mm
60mm
FKS10123682
100mm
12mm
110mm
2 Options
Catalog number
FKS10124352
Price (SGD)
136.00
PK
Add to cart
Diameter (Metric)Top:
55mm
Diameter (Metric) Stem:
8mm
Length (Metric) Stem:
60mm
Price (SGD)
136.00
PK
Add to cart
Manufactured from Soda Lime glass.
Short stem funnel
Less likely to clog when funneling thicker substances.
Specifications
Diameter (Metric)Top
55mm
Diameter (Metric) Stem
8mm
Material
Soda Lime Glass
Height (Metric)
95 mm
For Use With (Application)
Ideal for filtering applications in the laboratory
Length (Metric) Stem
60mm