Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Actual product may vary
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Fisherbrand
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Utilize these chemically inert, translucent and autoclavable funnels which feature a TFE stopcock assembly. Chemical-resistant, leakproof and requires no lubrication.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Includes
FKS101205512000 mL53 mm Closure
FIS10-437-25D1000 mL53 mm Closure
FIS10-437-25E2000 mL53 mm Closure
Catalog number FKS10120551
Price (SGD)
3,253.00
EA
Add to cart
Capacity (Metric):
2000 mL
Includes:
53 mm Closure
Price (SGD)
3,253.00
EA
Add to cart
Perform your liquid-liquid extractions with these chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer and Closure made with Tefzel™ which feature a TFE stopcock assembly. TFE stopcock assembly has excellent chemical resistance, is leakproof† when used with provided Nalgene closure and does not require lubrication — can be chemically disinfected. More transparent than our polypropylene separatory funnels. Compatible with all common lab reagents and solvents. Won't chip, crack or shatter like glass separatory funnels.
Features:
  • Break-resistant and impervious to any chemical used in a separatory funnel
  • No phthalates to contaminate analyses
  • Useful in radiological and hazardous separations
  • Autoclavable/Leakproof† when used with provided Nalgene closure
  • Stem length: 65mm
Ordering Information:
For replacement stopcock assembly for 125mL to 1L sizes, order Cat. No. 714301-1025. For replacement stopcock assembly for 2L size, order Cat. No. 714301-1050.
Note:
Funnel can be autoclaved; stopcock can be chemically sterilized. No stopcock lubrication needed.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)2000 mL
MaterialTeflon FEP
Height (Metric)401 mm
Includes53 mm Closure
Quantity1/Pk.; 2/Cs.
Product TypeSeparatory Funnel
Unit SizeEA