High-quality, low-cost analog polarimeter capable of measuring the rotation of optically active samples. Medline™ CETI 8kg Polaris Analog Polarimeter matches the performance of many digital instruments in the market today.
Have Questions?
Catalog number FKS10113944
Price (SGD)
10,454.00
EA
Add to cart
Price (SGD)
10,454.00
EA
Add to cart
Accepts sample tubes with lengths up to 220mm
Used for measuring concentrations and optical rotations in the food, sugar, flavouring, fragrance, pharmaceutical, and chemical industries
Supplied with two spare fuses, two test tubes (100 and 200mm), screwdriver, and instruction manual