Midisart™ 2000 PTFE Air Filter
Midisart™ 2000 PTFE Air Filter
Actual product may vary
Fisherbrand
Midisart™ 2000 PTFE Air Filter
Designed for easy handling and maximum safety in air filtration. Sartorius™ Midisart™ 2000 PTFE Air Filter entirely eliminates moisture breakthrough because of its inherently hydrophobic PTFE membrane reinforced with polypropylene gauze. Stepped hose barb connectors ensure a simple and secure hold.
Have Questions?
Catalog NumberQuantity
FKS1010966012Pack
Catalog number FKS10109660
Price (SGD)
712.00
PK
Add to cart
Quantity:
12Pack
Price (SGD)
712.00
PK
Add to cart
The Midisart™ filter is the clear choice for small-scale sterile air/gas filtration applications. Superior flow rates and seamless integration into single-use assemblies provide a distinct advantage that directly translates into cost savings.
  • 100% factory integrity tested to ensure inherent quality in every filter prior to use
  • Perfect traceability with optimized product labeling that includes lot and unit numbers
  • Lightweight (only 20g) so it will not weigh down or kink tubing
  • Can be sterilized by autoclaving at 134°C for up to 20 cycles
  • Applications include:
    • In-line sterilization of and particulate removal from air and gases, such as sterilization of air for small fermenters
    • Sterile venting of filling vessels and fermentation carboys, including culture vessels and CO2 incubators (6 to 120 liters)
    • Venting of holding tanks for sterile, distilled water and liquid culture media
    • Autoclave venting
Specifications
AutoclavableAutoclavable
Filter TypeMembrane Filter
FormatDisc
Material (Membrane)PTFE
Pore Size0.2 μm
SterilityNon-sterile
Filtration Area20 cm2
For Use With (Application)In-line sterilization of and particulate removal from air and gases and sterile venting of vessels, fermentation carboys, holding tanks and autoclaves
Housing MaterialPolypropylene
HydrophobicityHydrophobic
IncludesMultiple stepped hose barb connectors
Max. Inlet Pressure3Bar
Quantity12Pack
Temperature Range (Metric)134°C
TypeAir Filter
Diameter (Metric)64 mm
Unit SizePK