Pharmalyte™ Carrier Ampholytes
Pharmalyte™ Carrier Ampholytes
Actual product may vary
Fisher Catalogue
Pharmalyte™ Carrier Ampholytes
Use in Isoelectric focusing. Cytiva Pharmalyte™ Carrier Ampholytes prepared by the co-polymerization of glycine, glycylglycine, amines, and epichlorohydrin, are available in five broad-range and four narrow-range pH intervals.
Have Questions?
Catalog NumberpH Range
FKS101061545 to 8
Catalog number FKS10106154
View Price:Sign in to see your account pricing. Need an account? Register with us today.
pH Range:
5 to 8
  • Co-polymerisates of glycine, glycylglycine, amines and epichlorhydrin
  • Form extremely stable, linear pH gradients under the influence of electric fields
  • Available in both narrow and broad pH ranges to suit specific application
  • Broad ranges can be used directly for agarose, Ultrodex or polyacrylamide gels
  • Narrow ranges provide high resolution for dedicated applications using polyacrylamide gels
Specifications
ColorColorless to Light Yellow
For Use With (Equipment)PhastSystem Automated Electrophoresis Unit
pH Range5 to 8
Product TypePharmalyte carrier Ampholyte
FormLiquid
FormulationCo-polymerization of Glycine, Glycylglycine, Amines and Epichlorohydrin
For Use With (Application)IEF
Quantity25mL
Storage Requirements2° to 8°C
Unit SizeEA