Sealing Tape for 96-Well Plates
Sealing Tape for 96-Well Plates
Actual product may vary
Fisherbrand
Sealing Tape for 96-Well Plates
Clear, self-sticking sealing tape for standard microplates.
Have Questions?
Catalog NumberQuantity
FKS10105383100 sheets
Catalog number FKS10105383
Price (SGD)
-
Quantity:
100 sheets
Clear, self-sticking sealing tape for standard microplates.
Thermo Scientific Microplate Sealing Tape is a package of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well or 384-well microplates during incubation steps.
Features of Microplate Sealing Tape:
Pressure-sensitive—position the tape over plate and press to seal; easy to remove
Separate sheets—each sheet of tape is supplied on its own easy-to-peal backing (liner)
Durable and inert—film thickness provides support and has a thin layer of acrylic adhesive
Use the disposable sheets to seal and reseal a microplate two or three times for incubation steps in an ELISA or protein assay procedure. Seal plates quickly and easily to allow mixing or prevent evaporation during assay incubation steps.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorClear
MaterialPolyester
SterilityNon-sterile
No. of Wells96/384
Quantity100 sheets
Product TypeFilm, Seal, or Applicator
AdhesivePressure Sensitive
Plate Compatibility96/384-well Plate
Unit SizePK