PCR Plate, 96-well, non-skirted
PCR Plate, 96-well, non-skirted
Actual product may vary
Fisherbrand
PCR Plate, 96-well, non-skirted
96-well non-skirted plates for use in PCR and qPCR applications. Available with black lettering for improved sample tracking during pipetting.
Have Questions?
Catalog NumberQuantity
FKS1010084325 plates
Catalog number FKS10100843
Price (SGD)
352.00
PK
Add to cart
Quantity:
25 plates
Price (SGD)
352.00
PK
Add to cart
96-Well Non-Skirted Plates for use in PCR and qPCR applications hold up to 0.3 mL of well volume when paired with seals like adhesive films/foils, Thermo-Mat and cap strips. The plates feature a cut corner located at H1 and are compatible with most thermal cycles.
The well bottom is clear for maximum visibility. Available in assorted colors with clear or black alpha-numeric lettering. Compatible with standard multichannel pipettes.
The Thermo Scientific 96-Well Non-Skirted Plates for use in PCR and qPCR applications hold up to 0.3 mL of well volume when paired with seals like adhesive films/foils, Thermo-Mat and cap strips. The plates feature a cut corner located at H1 and are compatible with most thermal cycles.
The well bottom is clear for maximum visibility. Available in assorted colors with clear or black alpha-numeric lettering. Compatible with standard multichannel pipettes.
Highlights
  • Non-skirted format compatible with most thermal cyclers
  • Available with black alpha-numeric lettering
  • Cut corner: H1
  • Clear well bottom for sample visibility
  • 0.3 mL maximum well volume (when used with adhesive or heat seals)
  • Sealable using Thermo-Mat, Adhesive Films and Foils, Heat Seals and Flat and Domed Cap Strips
Applications
  • PCR
  • qPCR/Real-Time PCR
Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified RNase/DNase-free
Packaging25 Plate
ColorClear
Product TypePCR Microplate
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Skirt StyleNon-skirt
MaterialPolypropylene
Volume (Metric) Well0.3 mL
No. of Wells96
Volume (Metric) Working0.2 mL
Quantity25 plates
Reaction SpeedStandard
Unit SizePK