Check Order Status
LDPE Pasteur Pipettes
Actual product may vary
Fisherbrand
LDPE Pasteur Pipettes
LDPE Pasteur Pipettes are ideal for aliquots with very good reproducibility of the number of drops per milliliter.
Have Questions?
Change view
Catalog Number
Length (Metric)
Volume (Metric)
Diameter (Metric) Outer
Drops/mL
FKS10100531
152 mm
3 mL
2.8 mm
25 to 27
FKS10646341
152 mm
2 mL
3.3 mm
22 to 26
2 Options
Catalog number
FKS10100531
Price (SGD)
207.00
PK
Add to cart
Length (Metric):
152 mm
Volume (Metric):
3 mL
Diameter (Metric) Outer:
2.8 mm
Drops/mL:
25 to 27
Price (SGD)
207.00
PK
Add to cart
Manufactured from LDPE.
Very good reproducibility of the number of drops per milliliter.
Ideal for aliquots.
Pasteur pipettes can be filled and deep-frozen, or changed into a closed vessel by heat-sealing the tip.
The integrated pipetting bulb depresses easily, minimizing fatigue from frequent pipetting.
Resistant to gas or gamma radiation sterilization.
Specifications
Diameter (Metric) Outer
2.8 mm
Material
LDPE
Drops/mL
25 to 27
Quantity
500 Pack
Top Style
Bulb Shaped
Volume (Metric)
3 mL
Length (Metric)
152 mm
Type
Pasteur Pipet
Disposable
Single-use