1x1 image pixel for data collection
96 Well Round (U) Bottom Plate, TC Surface, No Lid, Pack of 1
96 Well Round (U) Bottom Plate, TC Surface, No Lid, Pack of 1
Actual product may vary
Fisherbrand
96 Well Round (U) Bottom Plate, TC Surface, No Lid, Pack of 1
Color:Clear; Sterility:Sterile; Lid:Without Lid
Have Questions?
Catalog NumberQuantity
FKS10099000Case of 50
Catalog number FKS10099000
Price (SGD)
497.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Quantity:
Case of 50
Price (SGD)
497.00
PK
Add to cart
Thermo Scientific™ Nunc MicroWell Plates with Nunclon Delta Surface are ideal in a wide variety of applications, including cloning experiments and tissue culture-based analytical systems. Thermo Scientific Nunc Cell Culture products offer solutions for cell growth and analysis and have been used by researchers worldwide since 1953. With a wide range of surface areas and treatments, we have products for all your basic or advanced cell-based research.
  • Stackable
  • Radiation sterilized
  • Condensation rings control evaporation and retain condensate
  • Keyed lids prevent accidental reversals
  • Dimensions: 86 × 128mm
Ordering Information:
Individually wrapped.
Specifications
BarcodeCustom Barcoding Available
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
CertifiedNunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
LidWithout Lid
Array8 x 12
Packaging10/Pack, 50/Case
Product LineNunc
ColorClear
Product TypeCell Culture Microplate
MaterialPolystyrene
Volume (Metric) Well300 μL
No. of Wells96
SterilitySterile
Surface TreatmentNunclon Delta
Well ShapeRound Bottom (U-well)
No. per Case50
No. per Pack1
QuantityCase of 50
Unit SizePK
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
178544Certificate of ComplianceDec 02, 2024FKS10099000
174141Certificate of ComplianceDec 02, 2024FKS10099000
166578Certificate of ComplianceDec 02, 2024FKS10099000
171198Certificate of ComplianceDec 02, 2024FKS10099000
181294Certificate of ComplianceDec 02, 2024FKS10099000
5 results displayed, search above for a specific certificate
Share catalog number, name or link