Whatman™ Kjeldahl Analysis Weighing Paper, Grade B-2
Whatman™ Kjeldahl Analysis Weighing Paper, Grade B-2
Actual product may vary
Fisherbrand
Whatman™ Kjeldahl Analysis Weighing Paper, Grade B-2
Dissolves residue-free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil. Whatman Kjeldahl Analysis Weighing Paper, Grade B-2 is thin, lightweight, and low in nitrogen for accurate chemical analysis.
Have Questions?
Catalog NumberDimensions (L x W)
FKS1009042476.2 x 76.2mm (3 x 3 in.)
Catalog number FKS10090424
Price (SGD)
183.00
PK
Add to cart
Dimensions (L x W):
76.2 x 76.2mm (3 x 3 in.)
Price (SGD)
183.00
PK
Add to cart
Dissolves residue-free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil. Whatman Kjeldahl Analysis Weighing Paper, Grade B-2 is thin, lightweight, and low in nitrogen for accurate chemical analysis.
  • Thin (0.04 mm) and lightweight (43 g/m2)
  • Transparent and smooth parchment surface
  • Simple weighing out and sample transfer
  • Reliable for quantitative transfer
  • Range of sheet sizes for ease-of-use
  • Low nitrogen paper that is residue-free for accurate chemical analysis
Specifications
FormatSheet
MaterialParchment Paper
GradeB-2
Quantity500 Pk.
Thickness (Metric)0.04 mm
Dimensions (L x W)76.2 x 76.2mm (3 x 3 in.)
Length (English)3 in.
Length (Metric)76.2 mm
Width (English)3 in.
Width (Metric)76.2 mm
Unit SizePK