Polypropylene Coplin Staining Jar
Polypropylene Coplin Staining Jar
Actual product may vary
Fisherbrand
Polypropylene Coplin Staining Jar
Improves on the classic tapered square design with a larger opening for easier placement and removal of slides. Bel Art™ Polypropylene Coplin Staining Jar makes working with slides easier.
Have Questions?
Catalog NumberSlide Count
FKS1008284410
Catalog number FKS10082844
Price (SGD)
26.00
EA
Add to cart
Slide Count:
10
Price (SGD)
26.00
EA
Add to cart
Improves on the classic tapered square design with a larger opening for easier placement and removal of slides. Bel Art™ Polypropylene Coplin Staining Jar makes working with slides easier.
    • Five internal slots hold up to 10 standard 25 x 75mm slides back to back
    • Steam autoclavable at 121ºC
    • Flat top
    • Easy grip knurled edge
    • Deep thread
    • Seal ring to prevent leaks
Specifications
AutoclavableAutoclavable
Bottom ShapeRound
Capacity (Metric)322 cm3
ColorWhite
DisposableReusable
MaterialPolypropylene
ShapeCylindrical
SterilityNon-sterile
Surface TreatmentNon-treated
TypeStaining Jar
QuantityEach
Slide Count10
Height (Metric)11.4 cm
Height (English)4.5 in.
Diameter (English)2.375 in.
Diameter (Metric)59.7 mm
Unit SizeEA