Overflow Measuring Flask
Overflow Measuring Flask
Actual product may vary
Fisherbrand
Overflow Measuring Flask
Aqualytic™ Overflow Measuring Flask is a suitable accessory for BD 600 and BD 606 respirometric method oxygen consumption measuring systems.
Have Questions?
Catalog NumberVolume (Metric)
FKS10079711428 mL
Catalog number FKS10079711
Price (SGD)
218.00
PK
Add to cart
Volume (Metric):
428 mL
Price (SGD)
218.00
PK
Add to cart
  • Suitable for BD 600 or BD 606 respirometric biochemical oxygen demand measuring system
  • Large volume flask
Specifications
DescriptionOverflow Flask
Volume (Metric)428 mL
Unit SizePK