Check Order Status
Porcelain Medium Form Melting Crucibles
Actual product may vary
Fisherbrand
Porcelain Medium Form Melting Crucibles
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.
Have Questions?
Catalog Number
Capacity (Metric)
FKS10077410
200 mL
Catalog number
FKS10077410
Price (SGD)
89.00
EA
Add to cart
Capacity (Metric):
200 mL
Price (SGD)
89.00
EA
Add to cart
Available in a variety of sizes
Sizes up to 175mL glazed except rim
1,000mL and larger are glazed except outside base
Complies with DIN VDE 0335, type C 100
Specifications
Material
Porcelain
Finish
Glazed Inside and Outside Except Base
Capacity (Metric)
200 mL
Diameter (Metric) Outer
80mm
Height (Metric)
65mm
Product Type
Melting Crucible