Nalgene™ Versi-Dry™ Surface Protectors
Nalgene™ Versi-Dry™ Surface Protectors
Actual product may vary
Nalgene™ Versi-Dry™ Surface Protectors
Nalgene™ Versi-Dry™ Surface Protectors
Nalgene™ Versi-Dry™ Surface Protectors
Nalgene™ Versi-Dry™ Surface Protectors
Nalgene™ Versi-Dry™ Surface Protectors
Nalgene™ Versi-Dry™ Surface Protectors
Fisher Catalogue
Nalgene™ Versi-Dry™ Surface Protectors
Quickly absorb spills and cushion breakable labware with these surface protectors. Use as a bench protector, shelf liner or cabinet liner to protect against the harshest of chemicals.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Description
FKS1007435018 in.Mat
62060-00
also known as CTA62060-00
300 ft.Roll
62080-0018 in.Mat
Catalog number FKS10074350
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
18 in.
Description:
Mat
Quickly absorb spills and cushion breakable labware with Thermo Scientific™ Nalgene™ Versi-Dry™ Surface Protectors. Each surface protector is composed of thousands of Thermo Scientific™ Thirsty Cells™ for heightened performance. The polyethylene backing protects the benchtop from the harshest of chemicals. Use as a bench protector, shelf liner, cabinet liner or for wiping up isolated spills.
  • Cushioned surface minimizes breakage of glass labware when dropped or tipped over
  • Skid resistant polyethylene backing resists harsh chemicals and protects your benchtop from spills and corrosion
  • Low-pile flat surface provides stability for glassware that is likely to tip
  • Versi-Dry quickly absorbs up to 750mL/m2 spills with its thousands of Thirsty Cells
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Product TypeSurface Protector
MaterialPolyethylene
Absorbancy750 mL/m2
Length (English)18 in.
Length (Metric)46 cm
Width (English)20 in.
Width (Metric)51 cm
DescriptionMat
No. per Case350
No. per Pack350
Quantity350 Pk.
Item DescriptionVersi-Dry Standard Absorbancy Mat 18 in. x 20 in. 350 mats/case
Unit SizePK