2000μL Polypropylene 96-Well Deep Well PCR Plates
2000μL Polypropylene 96-Well Deep Well PCR Plates
Actual product may vary
Fisherbrand
2000μL Polypropylene 96-Well Deep Well PCR Plates
Combine a new well design with a sophisticated color coding system for ease of reading and orientation.The well geometry of Eppendorf™ 2000μL Polypropylene 96-Well Deep Well PCR Plates allows high speed pipetting without splashing, excellent mixing and high speed centrifugation, which accelerates your manual and automated work processes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS10063712Yellow
FKS10245983White
FKS10675413Green
Catalog number FKS10063712
Price (SGD)
1,192.00
PK
Add to cart
Color:
Yellow
Price (SGD)
1,192.00
PK
Add to cart
  • Fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines.
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • LoBind - free of surface coating e.g., silicone
  • Free from DNA, DNase, RNase and PCR* inhibitor (PCR* clean)
  • LoBind Protein - minimum protein loss (less than 3% BSA 1μg/mL)
  • LoBind DNA - less than 1% loss of DNA/RNA at low concentrations and critical conditions (high salt, short DNA fragments)
  • Autoclavable
  • Optionally available with barcode
Specifications
FormatStandard
Array8 x 12
ColorYellow
MaterialPolypropylene
No. of Wells96
Total Volume (Metric) Well2000 μL
Unit SizePK