Chemical Duty Vacuum Pressure Pump
Chemical Duty Vacuum Pressure Pump
Actual product may vary
Fisherbrand
Chemical Duty Vacuum Pressure Pump
Decrease process filtration times. Chemical Duty Vacuum Pressure Pump supports high flow rates for faster pumping. Cast-aluminum pumphead, housing, and regulator with optional chemical-resistant internal components. Light weight and portable so they can be shared among workstations.
Have Questions?
Catalog NumberVoltageFrequency
FKS10052433220 V50 Hz
Catalog number FKS10052433
Price (SGD)
5,679.00
EA
Add to cart
Voltage:
220 V
Frequency:
50 Hz
Price (SGD)
5,679.00
EA
Add to cart
Decrease process filtration times. Chemical Duty Vacuum Pressure Pump supports high flow rates for faster pumping. Cast-aluminum pumphead, housing, and regulator with optional chemical-resistant internal components. Light weight and portable so they can be shared among workstations.
  • High output pumps feature a piston-driven design for greater power and high flow rates to decrease process filtration time
  • With optional chemical resistant head and diaphragm for use with corrosive chemicals, solvents and vapors
  • Portable design allows for easy sharing between workstations
  • Include a Millex™ FA50 filter for in-line moisture protection
  • UL listed and CE marked
Specifications
Max. Vacuum813 mbar (24 in.Hg)
Shipping Weight (English)9 lb.
Shipping Weight (Metric)4.1 kg
Includes70cm of 0.25 in. dia. tubing and Millex™ FA50 filter
Electrical Requirements220 V 50 Hz
MaterialAluminum
Weight (English)9 lb.
Weight (Metric)4.1 kg
Flow Rate37 L/min.
Frequency50 Hz
Overall Height (English)7 in.
Overall Height (Metric)17.8 cm
Overall Length (English)8 in.
Overall Length (Metric)20.3 cm
Pump TypePressure Pump
Maximum Pressure2.45 bar
Voltage220 V
Unit SizeEA