Nalgene™ Closed-Dome Polypropylene Tanks
Nalgene™ Closed-Dome Polypropylene Tanks
Actual product may vary
Fisherbrand
Nalgene™ Closed-Dome Polypropylene Tanks
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1005046175 L
FIS14-831-123210 L
FIS14-831-116378.5 L
Catalog number FKS10050461
Price (SGD)
3,226.00
EA
Add to cart
Capacity (Metric):
75 L
Price (SGD)
3,226.00
EA
Add to cart
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.
Features:
  • Tanks may be fitted with spigots
  • Autoclavable tanks and closures feature polypropylene construction
Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).
Warranty:
90 days
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceComply with FDA Reg. 177.1520 and USP Class VI
Diameter (English) Outer18.5 in.
Diameter (Metric) Outer47 cm
TypeClosed-dome Tank
MaterialPolypropylene
Capacity (English)20 gal.
Capacity (Metric)75 L
Height (English)35 in.
Height (Metric)81 cm
Unit SizeEA