Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Powder Funnel includes ground and fire-polished upper rim. Short, wide stem is optimal for funneling powder and other ground materials.
Have Questions?
Catalog Number
Diameter (Metric)Top
Diameter (Metric) Stem
FKS10049970
80 mm
22 mm
Catalog number FKS10049970
Price (SGD)
60.00
EA
Add to cart
Diameter (Metric)Top:
80 mm
Diameter (Metric) Stem:
22 mm
Price (SGD)
60.00
EA
Add to cart
Manufactured from borosilicate glass.
Ground and fire-polished upper rim.
Short, wide stem optimal for funneling powder and other ground materials.