Ice Flaker with Storage Bin
Ice Flaker with Storage Bin
Actual product may vary
Fisherbrand
Ice Flaker with Storage Bin
Extrudes ice flakes at a temperature just below zero degrees. Scotsman™ Ice Flaker with Storage bin can produce up to 70kg of flake per day and holds up to 25kg of storage. Flakes contain 25% residual water content and is stored inside an insulated, plastic lined containment bin with front opening for easy access.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1004978925 kg
Catalog number FKS10049789
Price (SGD)
18,228.00
EA
Add to cart
Capacity (Metric):
25 kg
Price (SGD)
18,228.00
EA
Add to cart
Ice flakes are extruded at a temperature just below zero degrees and contain 25% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
  • Up to 70kg (154 lbs.) of flake ice production per day
  • Up to 25kg (55 lbs.) built-in ice storage
  • Pressure-based water level monitoring system may work with demineralised water as well as with standard tap water
  • Electronically controlled operation, with self-diagnostic functions with external alarm lights providing operational information for early alert and fast diagnosis of operating issues
  • Plug-n-play configuration right out of the box - just connect incoming water line and water discharge
  • External components constructed from corrosion-resistant stainless steel, crafted for optimal aesthetic appeal through superior fit and finish
  • Ergonomically convenient, slanted & disappearing storage bin door
  • Front access for easier maintenance
  • Front-in, front-out airflow allows for built-in installations
  • CFC-free R-134a refrigerant
  • 120mm legs included, extensions available as an option Scotsman flaked ice is ideal for a variety of applications within laboratory and industrial environments.
Specifications
Certifications/ComplianceISO 9001
Capacity (Metric)25 kg
Depth (Metric)626mm
Product TypeAF80 Ice maker
For Use With (Application)May work with demineralised water as well as with standard water
Includes120mm legs
Hertz50Hz
MaterialCorrosion-resistant stainless steel
Height (Metric)813mm
Voltage230V
Width (Metric)535mm
Yield70kg/day
Electrical Requirements230V/50Hz
Warranty2 year manufacturer warranty
Unit SizeEA