Rack for Sedimentation Cones
Rack for Sedimentation Cones
Actual product may vary
Fisherbrand
Rack for Sedimentation Cones
Holds 2 Imhoff sedimentation cones made of plastic. Compact design and easy to carry, even with cones filled.
Have Questions?
Catalog NumberDimensions (L x W x H)
FKS10048850300 x 130 x 315 mm
Catalog number FKS10048850
Price (SGD)
448.00
PK
Add to cart
Dimensions (L x W x H):
300 x 130 x 315 mm
Price (SGD)
448.00
PK
Add to cart
  • Holds 2 Imhoff sedimentation cones
  • Polymethyl methacrylate (PMMA)/Polypropylene (PP) construction
Specifications
Product TypeImhoff Cone Rack
MaterialPMMA, Polypropylene
Dimensions (L x W x H)300 x 130 x 315 mm
Unit SizePK