Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Actual product may vary
Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Nalgene™ LDPE Economy Wash Bottles
Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS100483001000 mL38-430 mm
2401-0125125 mL24 mm
FIS03-409-10AA125 mL24 mm
2401-0250250 mL24 mm
FIS03-409-10BB250 mL24 mm
FIS03-409-10CC500 mL28 mm
2401-10001000 mL38-430 mm
FIS03-409-10DD1000 mL38-430 mm
Catalog number FKS10048300
Price (SGD)
99.00
PK
Add to cart
Capacity (Metric):
1000 mL
Closure Size:
38-430 mm
Price (SGD)
99.00
PK
Add to cart
High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
Features:
  • Polypropylene screw closure/stem with PPCO draw tube
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorTranslucent
IncludesStem and draw tube
MaterialLDPE
No. per Case12
No. per Pack4
Closure IncludedYes
GraduatedNo
MouthNarrow
ShapeCylindrical
Height (Metric)259 mm
Product TypeEconomy Wash Bottle
Height (English)10.2 in.
Closure MaterialPolypropylene
Closure Size38-430 mm
Diameter (Metric)72.6 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizePK