Porcelain Basins
Porcelain Basins
Actual product may vary
Fisherbrand
Porcelain Basins
Use these dishes for a variety of applications. Haldenwanger™ Porcelain Basins feature a smooth internal finish, constructed from optimized laboratory porcelain.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Length (Metric)
FKS1004816013 mm70 mm
FKS1003815016 mm96 mm
Catalog number FKS10048160
Price (SGD)
70.00
EA
Add to cart
Height (Metric):
13 mm
Length (Metric):
70 mm
Price (SGD)
70.00
EA
Add to cart
  • Shallow form, round-bottom porcelain basin
  • Available in a variety of sizes
  • Includes spout for easy pouring
  • Complies with DIN VDE 0335, type C 100
Specifications
MaterialPorcelain
ShapeRectangular
TypeEvaporating Basin
For Use With (Application)Laboratories
Interior FinishSmooth
Dimensions (L x W x H)70 x 46 x 13 mm
Length (Metric)70 mm
Height (Metric)13 mm
Unit SizeEA