Vertical Profile Auto-Desiccator Cabinet
Vertical Profile Auto-Desiccator Cabinet
Actual product may vary
Fisherbrand
Vertical Profile Auto-Desiccator Cabinet
Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with these Auto-Desiccator Cabinets
Have Questions?
Catalog NumberColor
FKS10046511Clear with Blue End Caps
Catalog number FKS10046511
Price (SGD)
6,060.00
EA
Add to cart
Color:
Clear with Blue End Caps
Price (SGD)
6,060.00
EA
Add to cart
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Automatic dehumidification and desiccant regeneration cycle operates continuously every 20 minutes
  • Color-indicating silica gel inside the desiccant regeneration module allows quick visual confirmation of desiccant status
  • Large door with easy-to-view digital hygrometer maximizes access to interior space; includes patented seal design, tab latches and security loops for a padlock or tamper-evident seals; opens right to left
  • Perforated shelves facilitate circulation of dry air
  • Compact 34.1W x 41.4cmD footprint for space-saving stacking up to 2 units high
Specifications
ColorClear with Blue End Caps
Cabinet StyleAuto-Desiccator
MaterialCopolyester
No. of Shelves3
No. per Pack1 Each
Shelf TypePerforated
Width (Metric)34.1 cm
Height (Metric)51.8 cm
Depth (Metric)41.4 cm