Check Order Status
Pasteur Pipettes
Actual product may vary
Fisherbrand
Pasteur Pipettes
Ideal for transferring samples to sample cups and for drop counting. Hirschmann Instruments™ Pasteur Pipettes are available in either long or short form.
Have Questions?
Catalog Number
Length (Metric)
FKS10044440
150 mm
Catalog number
FKS10044440
Price (SGD)
260.00
PK
Add to cart
Length (Metric):
150 mm
Price (SGD)
260.00
PK
Add to cart
Acc. to ISO 7712
Without cotton plug
With extended fine tip
Specifications
Certifications/Compliance
ISO 7712
Material
Glass
Includes
Finely tapered end
Length (Metric) Tip
40 mm
Quantity
1000 Pack
Top Style
Fine drawn
Volume (Metric)
2 mL
Length (Metric)
150 mm
Type
Pasteur Pipet