Telescopic Rod
Telescopic Rod
Actual product may vary
Telescopic Rod
Telescopic Rod
Fisherbrand
Telescopic Rod
Versatile sampling system with exchangeable tools for a wide range of applications.
Have Questions?
Catalog NumberLength (Metric)Includes
FKS10040822115 to 300 cm (Adjustable)3 Rods
Catalog number FKS10040822
Price (SGD)
283.00
EA
Add to cart
Length (Metric):
115 to 300 cm (Adjustable)
Includes:
3 Rods
Price (SGD)
283.00
EA
Add to cart
  • Different tools can be attached to the telescopic rod
  • The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint
  • Ideal for sampling from baths, tanks, shafts, open water courses etc
  • Sampling depths down to 6 m can be reached with the adjustable telescopic aluminum rod
  • The telescopic rods cannot be rotated radially, which means even better handling
  • The individual rods have been provided with an end stop, this means that one rod cannot accidentally slip out of another rod
  • Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm. Fits into every car boot!
  • Springs made of stainless steel
Specifications
Product TypeTelescopic Rod
MaterialAluminum
AutoclavableNot Autoclavable
Includes3 Rods
Tube MaterialAluminum
For Use With (Application)Industry and Water Treatment, Ideal for Sampling From Baths, Tanks, Shafts, Open Water Courses, Used with Telescope Sampler
For Use With (Equipment)Bürkle Angled beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net
Length (Metric)115 to 300 cm (Adjustable)
Unit SizeEA