1x1 image pixel for data collection
Quickfit™ Pressure Equalizing Funnel with Rotaflo™ Stopcock
Quickfit™ Pressure Equalizing Funnel with Rotaflo™ Stopcock
Actual product may vary
Fisherbrand
Quickfit™ Pressure Equalizing Funnel with Rotaflo™ Stopcock
For use when liquids have to be introduced into vessels under vacuum
Have Questions?
Catalog NumberCapacity (Metric)Cone SizeJoint Number
FKS10034804100 mL19/2619/26
Catalog number FKS10034804
Price (SGD)
753.00
EA
Add to cart
Capacity (Metric):
100 mL
Cone Size:
19/26
Joint Number:
19/26
Price (SGD)
753.00
EA
Add to cart
For use when liquids have to be introduced into vessels under vacuum
  • Complies with ISO 4800
  • Can also be used with Soxhlets as solvent traps to facilitate recovery at the end of an extraction
  • With GP Rotaflo™ stopcocks having chemically resistant PTFE keys
Specifications
Capacity (Metric)100 mL
MaterialBorosilicate Glass
Certifications/ComplianceISO 4800
Cone Size19/26
For Use With (Application)Used when liquids have to be introduced into vessels under vacuum, Soxhlets as solvent trap
Joint Number19/26
Quantity1PK
Stopcock3mm
Product TypePressure-equalizing Funnel
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link