ep Dualfilter T.I.P.S.™ Racks - Filter Pipette Tips
ep Dualfilter T.I.P.S.™ Racks - Filter Pipette Tips
Actual product may vary
Fisher Catalogue
ep Dualfilter T.I.P.S.™ Racks - Filter Pipette Tips
Eppendorf ep Dualfilter T.I.P.S.™ Racks offer 'PCR clean and Sterile' grade filter pipette tips, providing dual-layer protection against contamination for critical lab applications, ensuring precision, reliability, and hygiene in high-stakes environments
Have Questions?
Catalog NumberVolume (Metric)Clean ClaimsColor Code
FKS100292121000 to 10,000 μLPCR Clean and SterileTurquoise
Catalog number FKS10029212
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
1000 to 10,000 μL
Clean Claims:
PCR Clean and Sterile
Color Code:
Turquoise
Eppendorf™ ep Dualfilter T.I.P.S.™ Filter Tips offer ultimate pipette and sample protection against unintentional, and unnoticed contamination
  • Two filter layers provide dual protection against aerosols (1) and biomolecules to guard your pipette and sample
  • Free of PCR inhibiting additives provide for reliable downstream results
  • Available in PCR clean/Sterile (sterile and pyrogen-free)
  • Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
  • Lot-specific certificates are available
  • Certificate of filter efficiency acc. to EN 29463 available
  • Modern slim rack design with reclosable lid
  • Improved sustainability through 20 % lower polypropylene consumption per rack
  • ACT™-labeled by non-profit My Green Lab™ to help you make an informed product choice when it comes to sustainability
  • (1) An aerosol is a colloid of fine solid particles or liquid droplets, in air or another gas.
Specifications
Volume (Metric)1000 to 10,000 μL
FilteredFiltered
SterilitySterile
ColorColorless
Length (English)9.6 in.
Length (Metric)243 mm
Certifications/ComplianceEN 1822
Clean ClaimsPCR Clean and Sterile
For Use With (Application)DNA applications, RNA applications, Protein applications, Cell Culture applications
FormatRack
Packaging100 Tips, Individually Packed
Quantity100 Pk.
Product TypeFilter Pipette Tips
Color CodeTurquoise
Product Lineep DualFilter T.I.P.S