In-Line Filter Holder, 47 mm
In-Line Filter Holder, 47 mm
Actual product may vary
Fisherbrand
In-Line Filter Holder, 47 mm
Supports inline production of ultraclean liquids or gasses by pressure prefiltration
Have Questions?
Catalog NumberDiameter (Metric)
FKS1002464047 mm
Catalog number FKS10024640
Price (SGD)
1,728.00
EA
Add to cart
Diameter (Metric):
47 mm
Price (SGD)
1,728.00
EA
Add to cart
Supports inline production of ultraclean liquids or gasses by pressure prefiltration
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifications
AutoclavableNot Autoclavable
Material (Membrane)Polypropylene
SterilityNon-sterile
TypeLaboratory Filter Holders or Cyclones
Diameter (Metric)47 mm
Diameter (Metric) Filter7.6 cm
Diameter (Metric) Membrane7.6 cm
Unit SizeEA