8-channel Adapter
8-channel Adapter
Actual product may vary
Fisherbrand
8-channel Adapter
Use these adapters with VACUBOY aspiration tools. Integra Biosciences™ 8-Channel Adapters used with VACUBOY aspiration tools convert any vacuum source into a user-friendly aspiration system.
Have Questions?
Catalog NumberType
FKS100234518-Channel stainless steel adapter, 40mm
Catalog number FKS10023451
Price (SGD)
264.00
EA
Add to cart
Type:
8-Channel stainless steel adapter, 40mm
Price (SGD)
264.00
EA
Add to cart
  • 8-Channel adapter
  • Flame sterilizable for maximal safety
  • Fit a variety of tubes, flasks, bottles etc.
  • Accommodate 12- to 96-well plates
Can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • All parts in contact with fluids are autoclavable
Specifications
MaterialStainless Steel
Type8-Channel stainless steel adapter, 40mm
For Use With (Equipment)VACUBOY Aspiration Tools
Unit SizeEA