Disposable Flint Glass Tubes with Plain End
Disposable Flint Glass Tubes with Plain End
Actual product may vary
Fisherbrand
Disposable Flint Glass Tubes with Plain End
Specially annealed to reduce breakage along stress point
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)Quantity
FKS100222531 mL50 mm6 mm500/Tray
FIS14-958A1 mL50 mm6 mm500/Tray
FIS14-958C6 mL75 mm12 mm250/Tray
FIS14-958D10 mL100 mm13 mm250/Tray
FIS14-958G19 mL125 mm16 mm250/Tray
FIS14-958H23 mL150 mm16 mm250/Tray
Catalog number FKS10022253
Price (SGD)
175.00
PK
Add to cart
Capacity (Metric):
1 mL
Length (Metric):
50 mm
Diameter (Metric):
6 mm
Quantity:
500/Tray
Price (SGD)
175.00
PK
Add to cart
Specially annealed to reduce breakage along stress point
  • Round bottom
  • Smooth top and bottom to prevent cuts
Specifications
Capacity (Metric)1 mL
Length (Metric)50 mm
MaterialFlint Glass
DisposableSingle-use
For Use WithSpecially annealed to reduce breakage along stress point
Label TypeNone
PackagingTray
Quantity500/Tray
SterilityNon-sterile
Diameter (Metric)6 mm
Closure TypeKIM-KAP
Product TypeDisposable Culture Tube
Unit SizePK