RadEye™ G Series Personal Dose Rate Meters
RadEye™ G Series Personal Dose Rate Meters
Actual product may vary
Fisherbrand
RadEye™ G Series Personal Dose Rate Meters
Detect and measure even the smallest change in gamma and dose rate measurements with intrinsically safe Thermo Scientific™ RadEye™ G Series of personal radiation detectors.
Have Questions?
Catalog NumberDescription
FKS10016389RadEye G-10 with Red Label
Catalog number FKS10016389
Price (SGD)
10,325.00
EA
Add to cart
Description:
RadEye G-10 with Red Label
Price (SGD)
10,325.00
EA
Add to cart
Continuously monitor your radiation exposure in any nuclear or radiological emergency. The Thermo Scientific™ RadEye™ G Series of personal radiation detectors provide first responders with quick, reliable gamma and dose rate measurements. These light-weight, handheld Geiger counters detect and measure very low gamma energies; even the smallest change in radiation rates are displayed immediately, while coincidentally occurring fluctuations are suppressed. For hazmat teams investigating potentially explosive atmospheres, intrinsically safe RadEye G models meet the latest ATEX standards.
  • Menu driven interface can be optimized to application
  • Large, clear, backlit display for error free readings
  • High range versions for high turn back levels up to 300 R/h (3 Sv/h)
  • Intrinsically safe versions for potentially explosive environments
  • Large energy compensated GM tube for precise dose rate measurement of gamma and X-ray
  • High sensitivity to low energy gamma radiation in harsh environments
  • Durable – shock resistant design
  • 900 hour operation with 2 AAA batteries
  • Bright LED allows for operation in smoke and darkness
  • One hot and four advanced buttons – easy to use Alarm relay output – for area monitor application
  • Designed to exceed ANSI N42.33 standard
  • Intrinsically safe , ATEX certified
  • Data analysis can be done by an optional Windows™-based PC-software and an accompanying reader device
View our online training here.
Specifications
DescriptionRadEye G-10 with Red Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Dose Rate Meter
Measurement Range0.5 μSv/h to 100 mSv/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (66 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEA